Learn More
Waters Corp WATERS CORPORATION Leucine Enkephalin, Solid, 3 mg, MS Calibration
Catalog No. 501384606 Shop All Waters Chromatography Solutions Products
Encompass_Preferred
Leucine Enkephalin is a lyophilized peptide intended to be used as a reference material for mass spectrometer tuning and calibration. It helps ensure instruments are fine-tuned with precision, avoiding erroneous results caused by improperly adjusted parameters.
  • Provided as a lyophilized peptide for mass spectrometer tuning and calibration.
  • Each package contains two vials, each with 3 mg of solid leucine enkephalin.
  • Optimal storage at -20°C ± 10°C upon arrival.
  • Shelf life of 6 months after shipping.
  • Shelf life of 1 week upon reconstitution.
  • Suitable for use in various volumes, including 2 ng/μL, 1 ng/μL, 200 pg/μL, and 50 pg/μL.
  • Optimally used with a mixture of half acetonitrile and half water with 0.1% formic acid.
  • Easy to transfer into mixtures using a 1,000 μL pipette for precision.
Catalog No. 50-138-4606 Supplier Waters Corp Supplier No. 186006013
Leucine Enkephalin is a lyophilized peptide intended to be used as a reference material for mass spectrometer tuning and calibration. It helps ensure instruments are fine-tuned with precision, avoiding erroneous results caused by improperly adjusted parameters.
  • Provided as a lyophilized peptide for mass spectrometer tuning and calibration.
  • Each package contains two vials, each with 3 mg of solid leucine enkephalin.
  • Optimal storage at -20°C ± 10°C upon arrival.
  • Shelf life of 6 months after shipping.
  • Shelf life of 1 week upon reconstitution.
  • Suitable for use in various volumes, including 2 ng/μL, 1 ng/μL, 200 pg/μL, and 50 pg/μL.
  • Optimally used with a mixture of half acetonitrile and half water with 0.1% formic acid.
  • Easy to transfer into mixtures using a 1,000 μL pipette for precision.