Learn More
Cayman Chemical LeucInostatIn A 2.5mg
SDP
Catalog No. 502938409
Encompass_Preferred
A fungal metabolite with diverse biological activities; active against various fungi and bacteria in a disc assay; reduces TPA- and anti-CD3 antibody-induced T cell activation from 0.1-1 μg/ml; inhibits proliferation of DU145 prostate cancer cells co-cultured with PrSC stromal cells in vitro (IC50 = 0.045 μg/ml)
Catalog No. 50-293-8409 Supplier Cayman Chemical Supplier No. 281902.5 MG
A fungal metabolite with diverse biological activities; active against various fungi and bacteria in a disc assay; reduces TPA- and anti-CD3 antibody-induced T cell activation from 0.1-1 μg/ml; inhibits proliferation of DU145 prostate cancer cells co-cultured with PrSC stromal cells in vitro (IC50 = 0.045 μg/ml)