Learn More
Cayman Chemical Leucanicidin, Purity: ≥95%, CAS: 91021-66-8, 2.5 mg
SDP
Catalog No. 503015704
Encompass_Preferred
A macrolide bacterial metabolite; toxic to L. separata fourth instar larvae at 20 ppm; toxic to H. contortus, T. colubriformis, and O. circumcincta larvae (LD50s = 0.23-0.42 µg/ml)
Catalog No. 50-301-5704 Supplier Cayman Chemical Supplier No. 254792.5 MG
A macrolide bacterial metabolite; toxic to L. separata fourth instar larvae at 20 ppm; toxic to H. contortus, T. colubriformis, and O. circumcincta larvae (LD50s = 0.23-0.42 µg/ml)