Learn More
Medchemexpress LLC Leucocrystal violet | 603-48-5 | 98.39% | 10 G
SDP
Catalog No. 5000410234
Encompass_Preferred
Leucocrystal violet is a cationic triarylmethane dye that generates a colored purple product in the presence of hydrogen peroxide and horseradish peroxidase. It serves as a chromogenic reagent for the determination of submicrogram-level hydrogen peroxide without interference from tryptophan, persulfate, or chromate. It also reacts with blood to produce a purple/violet color in the presence of hydrogen peroxide and is widely used for staining blood residues on porous and non-porous materials.
  • Generates a colored purple product with hydrogen peroxide and horseradish peroxidase.
  • Functions as a chromogenic reagent for submicrogram hydrogen peroxide determination.
  • Reacts with blood to produce a purple/violet color.
  • Forms a weak fluorophore with specific absorption and emission wavelengths in unbuffered whole blood.
  • Used for staining blood residues on porous and non-porous materials.
Catalog No. 50-004-10234 Supplier Medchemexpress LLC Supplier No. HYD023310G
Leucocrystal violet is a cationic triarylmethane dye that generates a colored purple product in the presence of hydrogen peroxide and horseradish peroxidase. It serves as a chromogenic reagent for the determination of submicrogram-level hydrogen peroxide without interference from tryptophan, persulfate, or chromate. It also reacts with blood to produce a purple/violet color in the presence of hydrogen peroxide and is widely used for staining blood residues on porous and non-porous materials.
  • Generates a colored purple product with hydrogen peroxide and horseradish peroxidase.
  • Functions as a chromogenic reagent for submicrogram hydrogen peroxide determination.
  • Reacts with blood to produce a purple/violet color.
  • Forms a weak fluorophore with specific absorption and emission wavelengths in unbuffered whole blood.
  • Used for staining blood residues on porous and non-porous materials.