Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Leucocrystal violet, 99.8%, For HPLC analysis, MP Biomedicals™

Catalog No. ICN15519525 Shop All MP Biomedicals, Inc Products
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
ICN15519525 25 g
ICN15519505 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. ICN15519525 Supplier MP Biomedicals, Inc Supplier No. 0215519525
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

CAS: 603-48-5 | C25H31N3 | 373.54 g/mol

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 603-48-5
Molecular Formula C25H31N3
Molecular Weight (g/mol) 373.54
MDL Number MFCD00008314
InChI Key OAZWDJGLIYNYMU-UHFFFAOYSA-N
Synonym 4,4',4''-Methylidynetris(N,N-dimethylaniline)
PubChem CID 69048
IUPAC Name 4-{bis[4-(dimethylamino)phenyl]methyl}-N,N-dimethylaniline
SMILES CN(C)C1=CC=C(C=C1)C(C1=CC=C(C=C1)N(C)C)C1=CC=C(C=C1)N(C)C

Specifications

Color Gray/Purple
Quantity 25 g
Physical Form Crystalline Powder
Formula Weight 373.5
Percent Purity 99.8%
Grade Spectrophotometry
Chemical Name or Material Leucocrystal violet, For HPLC analysis
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.