Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC Leucocyanidin | 480-17-1 | 98.0% | 50 MG
SDP

Catalog No. 5000233274
Encompass_Preferred

Leucocyanidin is a plant-derived flavonoid research compound used in biochemical and pharmacological studies. It is supplied as a powder and in DMSO solution formats and is intended for research use only (not for human or clinical use).

  • High purity (98.0%).
  • Molecular formula C15H14O7 and molecular weight 306.27 g/mol.
  • Available as solid vials (5 mg-500 mg) and 10 mM 1 mL DMSO solutions.
  • Storage: powder -20°C (up to 3 years) or 4°C (up to 2 years); in solvent -80°C (6 months) or -20°C (1 month).
  • Soluble in DMSO.

Catalog No. 50-002-33274 Supplier Medchemexpress LLC Supplier No. HY11958050MG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Leucocyanidin is a plant-derived flavonoid research compound used in biochemical and pharmacological studies. It is supplied as a powder and in DMSO solution formats and is intended for research use only (not for human or clinical use).

  • High purity (98.0%).
  • Molecular formula C15H14O7 and molecular weight 306.27 g/mol.
  • Available as solid vials (5 mg-500 mg) and 10 mM 1 mL DMSO solutions.
  • Storage: powder -20°C (up to 3 years) or 4°C (up to 2 years); in solvent -80°C (6 months) or -20°C (1 month).
  • Soluble in DMSO.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.