Learn More
Medchemexpress LLC HY-119580 5mg , Leucocyanidin CAS:480-17-1 Purity:>98%
SDP
Supplier:  Medchemexpress LLC HY1195805MG
Encompass_Preferred
HY-119580 5mg Leucocyanidin CAS: 480-17-1 Leucocyanidin is an active anti-ulcerogenic ingredient was extracted from unripe plantain banana. Leucocyanidin demonstrates a significant protective effect against Aspirin-induced erosions in rat models. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-226-1689