Learn More
TARGETMOL CHEMICALS INC LEUCOMETHYLENE BLUE MESY 25MG
SDP
Catalog No. 502576369
Encompass_Preferred
Also available in 5 mg 10 mg 50 mg 100 mg 200 mg 1 mL 10 mM (in DMSO) and bulk. Please contact Fisher for quotes. Leucomethylene blue mesylate (TRx0237 mesylate) is a second-generation tau protein aggregation inhibitor for the treatment of Alzheimer's disease (AD) and frontotemporal dementia. purity: 99%
Catalog No. 50-257-6369 Supplier TARGETMOL CHEMICALS INC Supplier No. T700325MG
Also available in 5 mg 10 mg 50 mg 100 mg 200 mg 1 mL 10 mM (in DMSO) and bulk. Please contact Fisher for quotes. Leucomethylene blue mesylate (TRx0237 mesylate) is a second-generation tau protein aggregation inhibitor for the treatment of Alzheimer's disease (AD) and frontotemporal dementia. purity: 99%