Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC Leucomycin | 1392-21-8 | 200 MG
SDP

Catalog No. 5000419924
Encompass_Preferred

Leucomycin (Kitasamycin) is an orally active macrolide antibiotic derived from *Streptomyces kitasatoensis*. It exhibits broad-spectrum antibacterial activities against gram-positive and gram-negative bacteria, mycoplasma, leptospira, spirochaetes, rickettsiae, and some larger viruses.

  • Orally active macrolide antibiotic
  • Derived from *Streptomyces kitasatoensis*
  • Exhibits broad-spectrum antibacterial activities
  • Targets gram-positive and gram-negative bacteria, mycoplasma, leptospira, spirochaetes, rickettsiae, and some larger viruses
  • Appearance: solid
  • Color: white to off-white
  • In vitro solubility: 110 mg/mL in DMSO (requires ultrasonic); < 0.1 mg/mL in H2O (insoluble)
  • IC50 & Target: macrolide
  • Melting point: 128-145°C

Catalog No. 50-004-19924 Supplier Medchemexpress LLC Supplier No. HYN7112200MG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Leucomycin (Kitasamycin) is an orally active macrolide antibiotic derived from *Streptomyces kitasatoensis*. It exhibits broad-spectrum antibacterial activities against gram-positive and gram-negative bacteria, mycoplasma, leptospira, spirochaetes, rickettsiae, and some larger viruses.

  • Orally active macrolide antibiotic
  • Derived from *Streptomyces kitasatoensis*
  • Exhibits broad-spectrum antibacterial activities
  • Targets gram-positive and gram-negative bacteria, mycoplasma, leptospira, spirochaetes, rickettsiae, and some larger viruses
  • Appearance: solid
  • Color: white to off-white
  • In vitro solubility: 110 mg/mL in DMSO (requires ultrasonic); < 0.1 mg/mL in H2O (insoluble)
  • IC50 & Target: macrolide
  • Melting point: 128-145°C

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.