Learn More
AbD Serotec Leucoperm, 250 tests
Catalog No. NC1752617
Encompass
LEUCOPERM reagents allow intracellular antigen analysis with the same ease as surface antigens. The only prerequisite is the availability of suitable antibody conjugates. Most commercially available monoclonal antibody conjugates can be used with LEUCOPERM reagents. Some determinants are sensitive, however, to the fixation step involved. This and the optimal fixation time may have to be determined experimentally for each antibody conjugate.
Catalog No. NC1752617 Supplier AbD Serotec Supplier No. BUF09B
LEUCOPERM reagents allow intracellular antigen analysis with the same ease as surface antigens. The only prerequisite is the availability of suitable antibody conjugates. Most commercially available monoclonal antibody conjugates can be used with LEUCOPERM reagents. Some determinants are sensitive, however, to the fixation step involved. This and the optimal fixation time may have to be determined experimentally for each antibody conjugate.