Learn More
Selleck Chemical LLC Leucovorin (Folinic acid) Calcium Pentahydrate S1236-250mg
SDP
Catalog No. 503111288
Encompass_Preferred
Leucovorin (Folinic acid) Calcium Pentahydrate is a derivative of folic acid which can be used to increase levels of folic acid under conditions favoring folic acid inhibition Solutions are unstable and should be fresh-prepared
Catalog No. 50-311-1288 Supplier Selleck Chemical LLC Supplier No. S1236250MG
Add to Cart
Add to Cart
Leucovorin (Folinic acid) Calcium Pentahydrate is a derivative of folic acid which can be used to increase levels of folic acid under conditions favoring folic acid inhibition Solutions are unstable and should be fresh-prepared