Learn More
Apexbio Technology LLC Leucovorin Calcium, 25mg. Cas: 6035-45-6 MFCD: MFCD00149465. Derivative of folic acid
SDP
Supplier:  Apexbio Technology LLC A248925
Encompass_Preferred
Human lymphoid cell lines were studied for leucovorin requirements to protect from methotrexate (MTX)-induced growth suppression. Over a 72h continuous exposure leucovorin provided better protection to the cell lines LAZ-007 and RAJI than to the cell line Other sizes are available. Please inqury us for quote.
Catalog No. 50-190-4744