Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC Leucovorin-d4 (calcium hydrate) | >90.0% | 533.54 | C20H19D4CaN7O8 | 250 UG
SDP

Catalog No. 5000284065
Encompass_Preferred

Leucovorin-d4 (calcium hydrate) is a deuterium-labeled folinic acid supplied as the calcium hydrate salt for research and analytical applications. It is provided as a small research-grade standard (250 μg) with a listed purity of >90% and is not intended for human use.

  • Deuterium-labeled folinic acid for use as an isotope standard.
  • Calcium hydrate form suitable for analytical workflows.
  • Purity >90.0% as specified by the supplier.
  • Molecular formula C20H19D4CaN7O8 and molecular weight 533.54.
  • Supplied in a 250 μg research pack for analytical or mass spectrometry applications.
  • Not intended for human or clinical use; for research use only.

Catalog No. 50-002-84065 Supplier Medchemexpress LLC Supplier No. HYW653912A250UG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Leucovorin-d4 (calcium hydrate) is a deuterium-labeled folinic acid supplied as the calcium hydrate salt for research and analytical applications. It is provided as a small research-grade standard (250 μg) with a listed purity of >90% and is not intended for human use.

  • Deuterium-labeled folinic acid for use as an isotope standard.
  • Calcium hydrate form suitable for analytical workflows.
  • Purity >90.0% as specified by the supplier.
  • Molecular formula C20H19D4CaN7O8 and molecular weight 533.54.
  • Supplied in a 250 μg research pack for analytical or mass spectrometry applications.
  • Not intended for human or clinical use; for research use only.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.