Learn More
Cayman Chemical LeukadherIn 1 10mg
SDP
Catalog No. 502971854
Encompass_Preferred
An allosteric activator of CD11b/CD18; increases K562 cell adhesion to fibrinogen (EC50 = 4 µM); reduces rolling velocity of leukocytes, decreasing the number of leukocytes recruited to injured tissue; anti-inflammatory in a zebrafish tailfin injury model and a mouse model of nephritis; competitively inhibits YopH (Ki = 1.94 µM) and anthrax lethal factor metalloproteinase (IC50 = 6 µM)
Catalog No. 50-297-1854 Supplier Cayman Chemical Supplier No. 1485510 MG
An allosteric activator of CD11b/CD18; increases K562 cell adhesion to fibrinogen (EC50 = 4 µM); reduces rolling velocity of leukocytes, decreasing the number of leukocytes recruited to injured tissue; anti-inflammatory in a zebrafish tailfin injury model and a mouse model of nephritis; competitively inhibits YopH (Ki = 1.94 µM) and anthrax lethal factor metalloproteinase (IC50 = 6 µM)