Learn More
TARGETMOL CHEMICALS INC LEUKADHERIN-1 25MG
SDP
Catalog No. 502576901
Encompass_Preferred
Also available in 5 mg 10 mg 50 mg 100 mg 200 mg 1 mL 10 mM (in DMSO) and bulk. Please contact Fisher for quotes. Leukadherin-1 is an allosteric activator of CD11b/CD18. Increasing CD11b/CD18-dependent cell adhesion to fibrinogen Decreasing leukocyte motility and transendothelial migration; reduces inflammation. purity: 98%
Catalog No. 50-257-6901 Supplier TARGETMOL CHEMICALS INC Supplier No. T363825MG
Also available in 5 mg 10 mg 50 mg 100 mg 200 mg 1 mL 10 mM (in DMSO) and bulk. Please contact Fisher for quotes. Leukadherin-1 is an allosteric activator of CD11b/CD18. Increasing CD11b/CD18-dependent cell adhesion to fibrinogen Decreasing leukocyte motility and transendothelial migration; reduces inflammation. purity: 98%