Learn More
Discovery Life Sciences Leukemia ALL BMMC
SDP
Catalog No. 5000804123
Encompass
BMMCs Bone Marrow Mononuclear Cells are isolated from bone marrow aspirates and supplied as viably cryopreserved immune and progenitor cells commonly used in hematologic cancer research immune profiling and cell therapy development
Catalog No. 50-008-04123 Supplier Discovery Life Sciences Supplier No. 100320
Add to Cart
Add to Cart
BMMCs Bone Marrow Mononuclear Cells are isolated from bone marrow aspirates and supplied as viably cryopreserved immune and progenitor cells commonly used in hematologic cancer research immune profiling and cell therapy development