Learn More
Discovery Life Sciences Leukemia CLL BMMC
SDP
Catalog No. 5000804020
Encompass
BMMCs Bone Marrow Mononuclear Cells are isolated from bone marrow aspirates and supplied as viably cryopreserved immune and progenitor cells commonly used in hematologic cancer research immune profiling and cell therapy development
Catalog No. 50-008-04020 Supplier Discovery Life Sciences Supplier No. 100160
BMMCs Bone Marrow Mononuclear Cells are isolated from bone marrow aspirates and supplied as viably cryopreserved immune and progenitor cells commonly used in hematologic cancer research immune profiling and cell therapy development