Learn More
Discovery Life Sciences Leukemia CML Tx Naive PBMC
SDP
Catalog No. 5000804039
Encompass
PBMCs Peripheral Blood Mononuclear Cells are immune cells isolated from whole blood and viably cryopreserved providing a foundational material for immunology studies cell therapy research and functional immune assays
Catalog No. 50-008-04039 Supplier Discovery Life Sciences Supplier No. 121150
Add to Cart
Add to Cart
PBMCs Peripheral Blood Mononuclear Cells are immune cells isolated from whole blood and viably cryopreserved providing a foundational material for immunology studies cell therapy research and functional immune assays