Learn More
Creative Biomart Leukemia Inhibitory Factor (LIF)
Supplier:  Creative Biomart THP0264
Encompass_Preferred
Leukemia Inhibitory Factor (LIF) Human Recombinant produced in E.Coli is a single, non-glycosylated, polypeptide chain containing 180 amino acids and having a molecular mass of 19.7kDa. The Leukemia Inhibitory Factor (LIF) is purified by proprietary chromatographic techniques.
Catalog No. 50-208-4315