Learn More
Amsbio LLC Leukemia/Lymphoma Cancer Cell Pellet Array CPA07-01-5pk
Catalog No. 502041403
Encompass
FFPE cell line array sections manufactured using a proprietary technology that maintains both cell morphology and preserves nucleic acids and proteins. Includes cell lines JURKAT, CCRF-CEM, K562, RAJI, HUT78, SUDHL1, SUDHL4.
Catalog No. 50-204-1403 Supplier Amsbio LLC Supplier No. 30650120
FFPE cell line array sections manufactured using a proprietary technology that maintains both cell morphology and preserves nucleic acids and proteins. Includes cell lines JURKAT, CCRF-CEM, K562, RAJI, HUT78, SUDHL1, SUDHL4.