Learn More
Creative Biomart Leukocyte Elastase Inhibitor (LEI)
Catalog No. 502084316
Encompass_Preferred
Recombinant Leukocyte Elastase Inhibitor (LEI) Protein(His17-Glu332) is produced by E. coli expression system and fused with two N-terminal Tags, His-tag and T7-tag.
Catalog No. 50-208-4316 Supplier Creative Biomart Supplier No. THP0265
Recombinant Leukocyte Elastase Inhibitor (LEI) Protein(His17-Glu332) is produced by E. coli expression system and fused with two N-terminal Tags, His-tag and T7-tag.