Learn More
Bioworld LEUKOCYTE RNA PURIFICATION KIT
Catalog No. 50196919
Encompass_Preferred
For the rapid extraction and purification of total RNA from leukocytes Fractionate leukocytes from whole blood in minutes; Isolate total RNA, including microRNA, without phenol; Rapid and convenient spin-column format. FEATURES AND BENE...
Catalog No. 50-196-919 Supplier Bioworld Supplier No. 107600641
For the rapid extraction and purification of total RNA from leukocytes Fractionate leukocytes from whole blood in minutes; Isolate total RNA, including microRNA, without phenol; Rapid and convenient spin-column format. FEATURES AND BENE...