Learn More
AllCells LLC Leukopak- Cryopreserved- 2.5-3.0B Cells- (5 bags from the same donor)
Catalog No. NC2742219
Encompass
Leukopak- Cryopreserved- 2.5-3.0B CellsAll Cryopreserved Leukopaks are quality controlled for cell count and viability (pre-cryopreservation) using a validated AO/PI protocol. Additional characterization such as immunopheno-typing based on cell surface marker expression (CD45+- CD3+- CD4+- CD8+- CD14+- CD56+- CD19+) and 5-part differential CBC counts will be provided. This price reflects a 5 bag purchase (all from the same donor).
Catalog No. NC2742219 Supplier AllCells LLC Supplier No. LP CR SOLO 2.5 3.0B 5
Add to Cart
Add to Cart
Request a Quote
Leukopak- Cryopreserved- 2.5-3.0B CellsAll Cryopreserved Leukopaks are quality controlled for cell count and viability (pre-cryopreservation) using a validated AO/PI protocol. Additional characterization such as immunopheno-typing based on cell surface marker expression (CD45+- CD3+- CD4+- CD8+- CD14+- CD56+- CD19+) and 5-part differential CBC counts will be provided. This price reflects a 5 bag purchase (all from the same donor).