Learn More
AllCells LLC Leukopak, Cryopreserved, 2.5-3.0B Cells
Catalog No. 502067624
Encompass
Leukopak, Cryopreserved, 2.5-3.0B CellsAll Cryopreserved Leukopaks are quality controlled for cell count and viability (pre-cryopreservation) using a validated AO/PI protocol. Additional characterization such as immunopheno-typing based on cell surface marker expression (CD45+, CD3+, CD4+, CD8+, CD14+, CD56+, CD19+) and 5-part differential CBC counts will be provided.
Catalog No. 50-206-7624 Supplier AllCells LLC Supplier No. LP, CR, SOLO 2.53.0B
Add to Cart
Add to Cart
Leukopak, Cryopreserved, 2.5-3.0B CellsAll Cryopreserved Leukopaks are quality controlled for cell count and viability (pre-cryopreservation) using a validated AO/PI protocol. Additional characterization such as immunopheno-typing based on cell surface marker expression (CD45+, CD3+, CD4+, CD8+, CD14+, CD56+, CD19+) and 5-part differential CBC counts will be provided.