Learn More
Cayman Chemical Leukotriene A4 Hydrolase Polyclonal Antibody, 1 ea
SDP
Catalog No. 502968847
Encompass_Preferred
Immunogen: Recombinant human LTA4 hydrolase • Host: Rabbit • Species Reactivity: (+) Human; other species not tested • Applications: IHC and WB • Leukotriene A4 hydrolase is a bifunctional enzyme that converts leukotriene A4 to leukotriene B4 and acts as an aminopeptidase.
Catalog No. 50-296-8847 Supplier Cayman Chemical Supplier No. 1602501 EA
Immunogen: Recombinant human LTA4 hydrolase • Host: Rabbit • Species Reactivity: (+) Human; other species not tested • Applications: IHC and WB • Leukotriene A4 hydrolase is a bifunctional enzyme that converts leukotriene A4 to leukotriene B4 and acts as an aminopeptidase.