Learn More
Cayman Chemical LeukotrIen B4-d4 100ug
SDP
Catalog No. 502949588
Encompass_Preferred
An internal standard for the quantification of LTB4 by GC- or LC-MS.
Catalog No. 50-294-9588 Supplier Cayman Chemical Supplier No. 32011010 UG
An internal standard for the quantification of LTB4 by GC- or LC-MS.