Learn More
Cayman Chemical LeukotrIen B4-d5 10ug
SDP
Catalog No. 502911589
Encompass_Preferred
An internal standard for the quantification of LTB4 by GC- or LC-MS
Catalog No. 50-291-1589 Supplier Cayman Chemical Supplier No. 3609710 UG
An internal standard for the quantification of LTB4 by GC- or LC-MS