Learn More
Cayman Chemical LeukotrIen B4 dImthyl aMD 50ug
SDP
Catalog No. 502983783
Encompass_Preferred
LTB4 dimethyl amide is a moderate inhibitor of LTB4-induced degranulation of human neutrophils (Ki = 130 nM) and lysozyme release from rat PMNL.{379,384,1106} LTB4 dimethyl amide appears to be an antagonist of the LTB4 receptor on guinea pig lung membranes.{1106}
Catalog No. 50-298-3783 Supplier Cayman Chemical Supplier No. 2011550 UG
LTB4 dimethyl amide is a moderate inhibitor of LTB4-induced degranulation of human neutrophils (Ki = 130 nM) and lysozyme release from rat PMNL.{379,384,1106} LTB4 dimethyl amide appears to be an antagonist of the LTB4 receptor on guinea pig lung membranes.{1106}