Learn More
Cayman Chemical LeukotrIen B4 Pathway MaxSpe
SDP
Catalog No. 502995355
Encompass_Preferred
A mixture that contains 18-carboxy dinor leukotriene B4, 20-carboxy leukotriene B4, 20-hydroxy leukotriene B4, leukotriene B4, and 5(S)-HETE
Catalog No. 50-299-5355 Supplier Cayman Chemical Supplier No. 226401 EA
A mixture that contains 18-carboxy dinor leukotriene B4, 20-carboxy leukotriene B4, 20-hydroxy leukotriene B4, leukotriene B4, and 5(S)-HETE