Learn More
Cayman Chemical LeukotrIen B5 100ug
SDP
Catalog No. 502981163
Encompass_Preferred
A leukotriene; metabolite of EPA formed through 5-LO pathway; increases contraction of bullfrog lung strips ex vivo in a concentration-dependent manner; reduces tumor volume in mice injected with Tm1 murine melanoma cells (100 nM); elicits chemokinesis and lysosomal enzyme release from PMNLs; induces platelet aggregation
Catalog No. 50-298-1163 Supplier Cayman Chemical Supplier No. 21110100 UG
A leukotriene; metabolite of EPA formed through 5-LO pathway; increases contraction of bullfrog lung strips ex vivo in a concentration-dependent manner; reduces tumor volume in mice injected with Tm1 murine melanoma cells (100 nM); elicits chemokinesis and lysosomal enzyme release from PMNLs; induces platelet aggregation