Learn More
Cayman Chemical LeukotrIen D4 100ug
SDP
Catalog No. 502985484
Encompass_Preferred
An active metabolite of LTC4 and a constituent of SRS-A; induces contractions in isolated guinea pig parenchymal strips and tracheal spirals from 0.01-0.1 pM; increases insufflation pressure in anesthetized guinea pigs by 100% at 89 pmol/animal; intradermal administration increases vascular permeability in guinea pig skin at 0.1 pmol per animal
Catalog No. 50-298-5484 Supplier Cayman Chemical Supplier No. 20310100 UG
An active metabolite of LTC4 and a constituent of SRS-A; induces contractions in isolated guinea pig parenchymal strips and tracheal spirals from 0.01-0.1 pM; increases insufflation pressure in anesthetized guinea pigs by 100% at 89 pmol/animal; intradermal administration increases vascular permeability in guinea pig skin at 0.1 pmol per animal