Learn More
Cayman Chemical LeukotrIen E4 100ug
SDP
Catalog No. 502981157
Encompass_Preferred
Metabolite of LTD4 and one of the constituents of SRS-A; considerably less active (8 to 12-fold) than LTC4 in the biological activities characteristic of CysLTs; urinary excretion of LTE4 is often used as an indicator of asthma
Catalog No. 50-298-1157 Supplier Cayman Chemical Supplier No. 20410100 UG
Metabolite of LTD4 and one of the constituents of SRS-A; considerably less active (8 to 12-fold) than LTC4 in the biological activities characteristic of CysLTs; urinary excretion of LTE4 is often used as an indicator of asthma