Learn More
Cayman Chemical LeukotrIen F4 100ug
SDP
Catalog No. 502985099
Encompass_Preferred
LTF4 is a cysteinyl-leukotriene produced in vitro, but not reported to date in vivo. It is formed by the incubation of LTE4 with γ-glutamyl transpeptidase and glutathione. LTF4 is a weak agonist in its ability to contract vascular smooth muscle.{547} The rank order of potency of the cysteinyl-leukotrienes to contract vascular smooth muscle is LTD4 > LTC4 > LTE4 >> LTF4.{547,548}
Catalog No. 50-298-5099 Supplier Cayman Chemical Supplier No. 20520100 UG
LTF4 is a cysteinyl-leukotriene produced in vitro, but not reported to date in vivo. It is formed by the incubation of LTE4 with γ-glutamyl transpeptidase and glutathione. LTF4 is a weak agonist in its ability to contract vascular smooth muscle.{547} The rank order of potency of the cysteinyl-leukotrienes to contract vascular smooth muscle is LTD4 > LTC4 > LTE4 >> LTF4.{547,548}