Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC Leukotriene A4 Hydrolase Antibody (YA2400) | 50 UL
SDP

Catalog No. 5000444288
Encompass_Preferred

This Leukotriene A4 Hydrolase antibody (YA2400) is a rabbit-host antibody designed for laboratory chemical applications and substance manufacturing. It is reactive with human, mouse, and rat targets.

  • Host: Rabbit
  • Reactivity: Human, mouse, rat
  • Appearance: Liquid
  • Concentration: 0.3 mg/mL
  • Molecular weight: 69 kDa
  • Storage: Store at -20°C for 1 year; avoid repeated freeze/thaw cycles

Catalog No. 50-004-44288 Supplier Medchemexpress LLC Supplier No. HYP8265550UL
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

This Leukotriene A4 Hydrolase antibody (YA2400) is a rabbit-host antibody designed for laboratory chemical applications and substance manufacturing. It is reactive with human, mouse, and rat targets.

  • Host: Rabbit
  • Reactivity: Human, mouse, rat
  • Appearance: Liquid
  • Concentration: 0.3 mg/mL
  • Molecular weight: 69 kDa
  • Storage: Store at -20°C for 1 year; avoid repeated freeze/thaw cycles

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.