Learn More
Cayman Chemical Leukotriene A4d5 methyl ester
SDP
Catalog No. 502889437
Encompass_Preferred
An internal standard for the quantification of LTA4 methyl ester by GC- or LC-MS
Catalog No. 50-288-9437 Supplier Cayman Chemical Supplier No. 10006197100 UG
Add to Cart
Add to Cart
An internal standard for the quantification of LTA4 methyl ester by GC- or LC-MS