Learn More
Cayman Chemical Leukotriene A4d5 methyl ester
SDP
Supplier:  Cayman Chemical 1000619725 UG
Encompass_Preferred
An internal standard for the quantification of LTA4 methyl ester by GC- or LC-MS
Catalog No. 50-289-0949