Learn More
Cayman Chemical Leukotriene A4d5 methyl ester
SDP
Catalog No. 502887930
Encompass_Preferred
An internal standard for the quantification of LTA4 methyl ester by GC- or LC-MS
Catalog No. 50-288-7930 Supplier Cayman Chemical Supplier No. 1000619750 UG
An internal standard for the quantification of LTA4 methyl ester by GC- or LC-MS