Learn More
Eurofins Discoverx Leukotriene B4, 10 μg. (92-1171)
Supplier:  Eurofins Discoverx 921171
Encompass
Leukotriene B4 is a pro-inflammatory dihydroxy fatty acid derived from arachidonic acid and produced from leukocytes in response to inflammation. Agonist of the BLT1 and BLT2 receptors. Promotes adhesion and activation of leukocytes at the endothelium, and stimulates production of pro-inflammatory cytokines and mediators. May be a factor in inflammatory diseases, and shows antiviral activity towards DNA viruses and retroviruses. Provided as 10 μg of dry powder. These highly purified, pre-validated control ligands have been used to qualify our InCELL Hunter,  PathHunter and cAMP Hunter cell lines as well as our Assay Ready Kits. Ligands are conveniently supplied with a recommended reconstitution buffer. For detailed information and product-specific datasheet, visit www.discoverx.com/ligands.
Catalog No. 50-199-9699