Learn More
Cayman Chemical Leukotriene B4-3-aminopropyla
SDP
Catalog No. 502982881
Encompass_Preferred
An analog of LTB4 that binds to the BLT1 and BLT2 receptors with Ki values of 5.1 and 1,227 nM, respectively
Catalog No. 50-298-2881 Supplier Cayman Chemical Supplier No. 2011450 UG
An analog of LTB4 that binds to the BLT1 and BLT2 receptors with Ki values of 5.1 and 1,227 nM, respectively