Learn More
Cayman Chemical Leukotriene B4-3-aminopropyla
SDP
Catalog No. 502984710
Encompass_Preferred
An analog of LTB4 that binds to the BLT1 and BLT2 receptors with Ki values of 5.1 and 1,227 nM, respectively
Catalog No. 50-298-4710 Supplier Cayman Chemical Supplier No. 2011425 UG
Add to Cart
Add to Cart
An analog of LTB4 that binds to the BLT1 and BLT2 receptors with Ki values of 5.1 and 1,227 nM, respectively