Learn More
Medchemexpress LLC Leukotriene B4 | 71160-24-2 | MFCD00135630 | >=97.0% | 336.5 g/mol | C20H32O4 | 25 UG
SDP
Catalog No. 5000240422
Encompass_Preferred
Leukotriene B4 is a bioactive lipid mediator that acts as a potent chemoattractant and activator of leukocytes. It is used in research to investigate inflammation, leukocyte chemotaxis, receptor signaling, and related pharmacological responses, and can be employed as a synthetic intermediate in chemical biology applications.
  • Potent chemoattractant for leukocyte activation and migration.
  • Used in in vitro assays of inflammation and receptor signaling.
  • Suitable for pharmacology and signaling studies involving BLT receptors.
  • Research-grade reagent with reported purity ≥97%.
  • Well-defined chemical identifiers for reproducible research (CAS 71160-24-2, C20H32O4, MW 336.5 g/mol).
Catalog No. 50-002-40422 Supplier Medchemexpress LLC Supplier No. HY10760825UG
Add to Cart
Add to Cart
Leukotriene B4 is a bioactive lipid mediator that acts as a potent chemoattractant and activator of leukocytes. It is used in research to investigate inflammation, leukocyte chemotaxis, receptor signaling, and related pharmacological responses, and can be employed as a synthetic intermediate in chemical biology applications.
  • Potent chemoattractant for leukocyte activation and migration.
  • Used in in vitro assays of inflammation and receptor signaling.
  • Suitable for pharmacology and signaling studies involving BLT receptors.
  • Research-grade reagent with reported purity ≥97%.
  • Well-defined chemical identifiers for reproducible research (CAS 71160-24-2, C20H32O4, MW 336.5 g/mol).