Learn More
Medchemexpress LLC Leukotriene B4-d4 25ug | 124629-74-9 | 25UG
SDP
Catalog No. 5000240847
Encompass_Preferred
Leukotriene B4-d4 is the deuterium labeled Leukotriene B4 Leukotriene B4 (LTB4) is known as one of the most potent chemoattractants and activators of leukocytes and is involved in inflammatory diseases Leukotriene B4 is also an alkyl chain-based PROTAC linker that can be used in the synthesis of PROTACs[1 [2 [3
Catalog No. 50-002-40847 Supplier Medchemexpress LLC Supplier No. HY107608S25UG
Leukotriene B4-d4 is the deuterium labeled Leukotriene B4 Leukotriene B4 (LTB4) is known as one of the most potent chemoattractants and activators of leukocytes and is involved in inflammatory diseases Leukotriene B4 is also an alkyl chain-based PROTAC linker that can be used in the synthesis of PROTACs[1 [2 [3