Learn More
Cayman Chemical Leukotriene B4-d4, 124629-74-9, 25 g
SDP
Supplier:  Cayman Chemical 32011025
Encompass_Preferred
Molecular Formula C20H28D4O4, Purity ≥99% deuterated forms (d1-d4), Formula Weight 340.5, Synonyms: LTB4-d4; An internal standard for the quantification of LTB4 by GC- or LC-MS.
Catalog No. NC9799588