Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ Multispecies Leukotriene B4 Competitive ELISA Kit

Catalog No. ENEHLTB4
Encompass_Preferred
Change view
Click to view available options
Quantity:
96 Tests
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
ENEHLTB4 96 Tests
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. ENEHLTB4 Supplier Invitrogen™ Supplier No. EHLTB4
Add to Cart
Add to Cart

ELISA

The Multispecies Leukotriene B4 (LTB4) ELISA quantitates LTB4 in human saliva, urine, porcine plasma and tissue culture medium. Principle of the method This LTB4 competitive ELISA kit is an immunoassay for the quantitative determination of leukotriene B4 (LTB4) in biological fluids. The assay is based on the competition between LTB4 in the standard or sample and alkaline phosphatase conjugated LTB4 (LTB4-AP) for a limited amount of LTB4 monoclonal antibody bound to an anti-Rabbit IgG precoated 96-well plate. As the concentration of LTB4 in the sample increases, the amount of LTB4-AP captured by the coating antibody decreases. Thus, there is an inverse relationship between optical density (OD) and the amount of analyte in the sample. Rigorous validation Each manufactured lot of this ELISA kit is quality tested for criteria such as sensitivity, specificity, precision, and lot-to-lot consistency. See manual for more information on validation.

Leukotriene B4 (LTB4) is a leukotriene involved in inflammation. It is produced from leukocytes in response to inflammatory mediators and is able to induce the adhesion and activation of leukocytes on the endothelium, allowing them to bind and cross into the tissue. In neutrophils, LTB4 is a potent chemoattractant, and is able to induce the formation of reactive oxygen species and the release of lysosomal enzymes.
TRUSTED_SUSTAINABILITY

Specifications

Assay Range 11.7-120000 pg/mL
Assay Sensitivity <5.63 pg/mL
Content And Storage Sufficient For: 96 microplate wells
• Anti-Rabbit IgG Precoated 96-well Plate, 1 each
• Reagent Diluent, 30 mL
• LTB4 Antibody, 6 mL
• LTB4-AP Conjugate, 6 mL
• LTB4 Standard, 0.5 mL
• 10X Wash Buffer, 30 mL
• Substrate Solution, 20 mL
• Stop Solution, 6 mL
• Plate Sealer, 1 each
• LTB4 Assay Layout Sheet, 1 each
Description LTB4 Competitive ELISA Kit
Conjugate Alkaline Phosphatase (AP)
Immunoassay Kit Format Sandwich ELISA Kit
Product Type ELISA
Protein Family Cytokines & Receptors
Sample Type Plasma, Saliva, Supernatant, Urine
Specificity No significant cross-reaction with PGE2, PGF2a, 20-OH-LTB4, 20-COOH-LTB4, LTC4, LTD4, LTE4, 5(S)-HETE, 12(S)-HETE, 15(S)-HETE; less than 1% cross-reaction with 12-epi-LTB4; approx. 5% cross-reaction with 6-trans-LTB4 and 6-trans-12-epi-LTB4
Sufficient For 96 microplate wells
Target Species Human, Pig
For Use With (Equipment) Absorbance Microplate Reader
Gene Alias BLT; BLT1; BLTR; Chemoattractant receptor-like 1; chemokine receptor-like 1; CMKRL1; G protein-coupled receptor 16; GPR16; G-protein coupled receptor 16; leukotriene B4 receptor; leukotriene B4 receptor 1; LTB4R; LTB4-R 1; LTB4R1; LTB4-R1; LTBR1; P2RY7; P2Y purinoceptor 7; P2Y7; purinergic receptor P2Y, G-protein coupled, 7BLT; BLT1; BLTR; Chemoattractant receptor-like 1; chemokine receptor-like 1; CMKRL1; G protein-coupled receptor 16; GPR16; G-protein coupled receptor 16; leukotriene B4 receptor; leukotriene B4 receptor 1; LTB4R; LTB4-R 1; LTB4R1; LTB4-R1; LTBR1; P2RY7; P2Y purinoceptor 7; P2Y7; purinergic receptor P2Y, G-protein coupled, 7;
Interassay CV 12.4%
Intraassay CV 6.5%
Kit Contents Pre-coated 96 well plate, Standard, Assay Diluent, Detection Antibody, AP Conjugate, Wash Buffer, Chromogen, Stop Solution, Adhesive Plate Covers, Standard, Assay Diluent, Detection Antibody, AP Conjugate, Wash Buffer, Chromogen, Stop Solution, Adhesive Plate Covers
Label or Dye AP
Measurement Absorbance at 405 nm minus absorbance (at 570 nm)
Precision Dependent upon sample type and level
Protein Aliases LTB4; C20H32O4
Quantity 96 Tests
Regulatory Status RUO
Sample Volume Plasma 50 μL, Saliva 25 μL, Supernatant 100 μL, Urine 100 μL
Storage Requirements -20°C
Target Leukotriene B4
Test Time 1 hr 20 min
Total Assay Time 4 hr
Show More Show Less
WARNING: Reproductive Harm - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.