Learn More
Cayman Chemical Leukotriene C4, 72025-60-6, 50 g
SDP
Supplier:  Cayman Chemical 2021050
Encompass_Preferred
Molecular Formula C30H47N3O9S, Purity ≥97%, Formula Weight 625.8, Synonyms: LTC4; The parent CysLT produced by the LTC4 synthase-catalyzed conjugation of glutathione to LTA4; potent inducer of bronchoconstriction and enhanced vascular permeability that contributes to the pathogenesis of asthma and acute allergic hypersensitivity.
Catalog No. NC1332554