Learn More
Medchemexpress LLC Leukotriene C4 | 72025-60-6 | ≥97.0% | 625.77 | 25 UG
SDP
Catalog No. 5000439248
Encompass_Preferred
Leukotriene C4 is an eicosanoid lipid mediator produced by immune cells during type 2 inflammation. It can mediate inflammation, allergy, bronchoconstriction, and vascular leakage. It is also the parent cysteinyl leukotriene, produced by the LTC4 synthase catalyzed conjugation of glutathione to LTA4. Leukotriene C4 is produced by neutrophils, macrophages, mast cells, and by transcellular metabolism in platelets. It is one of the constituents of slow-reacting substance of anaphylaxis (SRS-A) and exhibits potent smooth muscle contracting activity.
  • Endogenous metabolite
  • Inflammation mediator
  • Research use only
  • High purity
  • Multiple formulations
  • Storage stability
Catalog No. 50-004-39248 Supplier Medchemexpress LLC Supplier No. 25UGHY113446
Leukotriene C4 is an eicosanoid lipid mediator produced by immune cells during type 2 inflammation. It can mediate inflammation, allergy, bronchoconstriction, and vascular leakage. It is also the parent cysteinyl leukotriene, produced by the LTC4 synthase catalyzed conjugation of glutathione to LTA4. Leukotriene C4 is produced by neutrophils, macrophages, mast cells, and by transcellular metabolism in platelets. It is one of the constituents of slow-reacting substance of anaphylaxis (SRS-A) and exhibits potent smooth muscle contracting activity.
  • Endogenous metabolite
  • Inflammation mediator
  • Research use only
  • High purity
  • Multiple formulations
  • Storage stability