Learn More
Medchemexpress LLC Leukotriene C4-d5 | 1441421-73-3 | 97.9% | 630.80 | 25 UG
SDP
Catalog No. 5000439446
Encompass_Preferred
Leukotriene C4-d5 is a deuterium-labeled version of Leukotriene C4. Leukotriene C4 is a cysteinyl leukotriene, which is formed when LTC4 synthase catalyzes the conjugation of glutathione to LTA4. This product is for research use only.
  • Deuterium labeled Leukotriene C4.
  • Can be used as a tracer.
  • Can be used as an internal standard for quantitative analysis by NMR, GC-MS, or LC-MS.
Catalog No. 50-004-39446 Supplier Medchemexpress LLC Supplier No. 25UGHY113446S
Leukotriene C4-d5 is a deuterium-labeled version of Leukotriene C4. Leukotriene C4 is a cysteinyl leukotriene, which is formed when LTC4 synthase catalyzes the conjugation of glutathione to LTA4. This product is for research use only.
  • Deuterium labeled Leukotriene C4.
  • Can be used as a tracer.
  • Can be used as an internal standard for quantitative analysis by NMR, GC-MS, or LC-MS.