Learn More
Cayman Chemical Leukotriene C4 methyl ester
SDP
Catalog No. 502887503
Encompass_Preferred
A more lipid soluble formulation of LTE4
Catalog No. 50-288-7503 Supplier Cayman Chemical Supplier No. 1000716450 UG
A more lipid soluble formulation of LTE4