Learn More
Cayman Chemical Leukotriene C4d5
SDP
Catalog No. 502890981
Encompass_Preferred
An internal standard for the quantification of LTC4 by GC- or LC-MS
Catalog No. 50-289-0981 Supplier Cayman Chemical Supplier No. 10006198100 UG
An internal standard for the quantification of LTC4 by GC- or LC-MS