Learn More
Enzo Life Sciences Leukotriene D4 (50µg). CAS: 73836-78-9
SDP
Supplier:  Enzo Life Sciences BMLLD0040050
Encompass_Preferred
Potent agonist for contraction of smooth muscle. Induces microvascular permeability and mucus secretion in airways. Enhances growth of human airway epithelial cells. Reduces myocardial contractility and coronary blood flow. Induces long lasting hypotension. Biosynthesis from LTC4 by γ-glutamyltranspeptidase. Alternative name: LTD4. Formulation: Solution in methanol/ammonium acetate buffer, 70:30, pH 5.6. Purity: ≥98% (HPLC). Long Term Storage: -80°C.
Catalog No. 50-201-1145